Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Dual Monochromator, 200-1000 nm
Catalog No. 5000014376
Encompass_Preferred
The SpectraMax M2 system is a dual-mode multi-detection microplate reader bundled with SoftMax Pro software for data acquisition and analysis. It provides flexible absorbance and fluorescence measurements with tunable monochromators for assay development.
  • Multi-mode detection: absorbance and fluorescence measurements.
  • Dual monochromators with 1 nm wavelength resolution.
  • Wavelength range: absorbance 200-1000 nm; fluorescence excitation 250-850 nm; emission 360-850 nm.
  • Supports 6, 12, 24, 48, 96, and 384 well plates.
  • Dual-mode cuvette port for cuvette measurements.
  • Top-reading capability for fluorescence assays.
  • Compact footprint: 8.6 x 22.8 x 15 in; weight 35 lb.
  • Compatible with software for acquisition, analysis, and reporting.
Catalog No. 50-000-14376 Supplier Molecular Devices Supplier No. M2
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M2 system is a dual-mode multi-detection microplate reader bundled with SoftMax Pro software for data acquisition and analysis. It provides flexible absorbance and fluorescence measurements with tunable monochromators for assay development.
  • Multi-mode detection: absorbance and fluorescence measurements.
  • Dual monochromators with 1 nm wavelength resolution.
  • Wavelength range: absorbance 200-1000 nm; fluorescence excitation 250-850 nm; emission 360-850 nm.
  • Supports 6, 12, 24, 48, 96, and 384 well plates.
  • Dual-mode cuvette port for cuvette measurements.
  • Top-reading capability for fluorescence assays.
  • Compact footprint: 8.6 x 22.8 x 15 in; weight 35 lb.
  • Compatible with software for acquisition, analysis, and reporting.