Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices CloneMedia CHO Growth A, Semi-solid, Without L-Gln

Catalog No. 5000014646
Encompass_Preferred

Viscous semi-solid medium formulated to immobilize single CHO cells and support clonal colony formation for cloning, screening, and automated picking workflows. Supplied without L-glutamine to enable controlled supplementation and compatible with ClonePix workflows.

  • Viscous semi-solid formulation that immobilizes single cells for distinct colony formation.
  • Supplied without L-glutamine to allow controlled addition of glutamine during workflows.
  • Contains methylcellulose, recombinant proteins, and supplements to support colony growth.
  • Suitable for suspension-adapted CHO, DG44, and DUXB11 cell lines.
  • Format: 1 x 90 mL vial; store at -20°C, thaw and use within one week at 2-8°C.

Catalog No. 50-000-14646 Supplier Molecular Devices Supplier No. K8840
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Viscous semi-solid medium formulated to immobilize single CHO cells and support clonal colony formation for cloning, screening, and automated picking workflows. Supplied without L-glutamine to enable controlled supplementation and compatible with ClonePix workflows.

  • Viscous semi-solid formulation that immobilizes single cells for distinct colony formation.
  • Supplied without L-glutamine to allow controlled addition of glutamine during workflows.
  • Contains methylcellulose, recombinant proteins, and supplements to support colony growth.
  • Suitable for suspension-adapted CHO, DG44, and DUXB11 cell lines.
  • Format: 1 x 90 mL vial; store at -20°C, thaw and use within one week at 2-8°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.