Learn More
Cygnus Technologies CHO HCP Antigen Concentrate, 3G
Supplier:  Cygnus Technologies F553H1
Encompass
Positive control for spike and recovery testing in ELISA. Actual HCP potency may change with age. Establish the precise value by comparison to kit standards in the CHO HCP ELISA Kit (F550-1). Concentration 1.5 mg/ml. This item is not returnable.
Catalog No. NC1766610