Learn More
Abcam CHO HCP ELISA kit (Host cell protein), 96 Tests
Description
- Assay Type: Sandwich
- Detection Method: Colorimetric
- Species Reactivity: Species independent
- Sensitivity: 10 ng/mL
- Range: 3.3 - 810 ng/mL
- Assay Time: 3 hr
- Results Type: Quantitative
- Storage: Store at +4°C
Specifications
Specifications
| Assay Range | 3.3 to 810 ng/mL |
| Assay Sensitivity | 10 ng/mL |
| Content And Storage | Store at 4°C |
| Detection Method | Colorimetric |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA Kit |
| Target Species | Non-species Specific |
| For Use With (Application) | ELISA |
| Quantity | 96 Tests |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.