Learn More
Enzo Life Sciences CHO host cell proteins polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2600100
Encompass_Preferred
Immunogen: CHO host cell proteins. Species reactivity: Hamster. Specificity: Recognizes CHO host cell proteins. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 10ppm Kathon.
Catalog No. 50-201-2805