Learn More
ACROBiosystems CHO/Human IL-1 RAcP Stable Cell Line (CCHO-ATP257) - 2 vials, ≥5 x 10^6 cells per vial, frozen
Catalog No. NC3900408
Encompass_Preferred
CHO/Human IL-1 RAcP Stable Cell Line is an adherent CHO-derived cell line engineered to express full-length human IL-1 RAcP. Supplied frozen in cryopreserved vials, the line is QC-verified for surface expression and validated for passage stability to support reproducible cell-based binding assays and bioassay development.
  • Provided as frozen vials with ≥5 x 10^6 cells per vial.
  • Surface expression confirmed by flow cytometry.
  • Suited for cell-based IL-1 RAcP binding and bioassay development.
  • Adherent CHO cell line with Zeocin selection (200 μg/mL).
  • Mycoplasma- and sterility-tested negative.
  • Validated passage stability and full traceable QC records.
  • Growth medium: F-12K + 10% FBS; freeze medium: serum-free cryopreservation medium.
Catalog No. NC3900408 Supplier ACROBiosystems Supplier No. CCHOATP2572VIALS
CHO/Human IL-1 RAcP Stable Cell Line is an adherent CHO-derived cell line engineered to express full-length human IL-1 RAcP. Supplied frozen in cryopreserved vials, the line is QC-verified for surface expression and validated for passage stability to support reproducible cell-based binding assays and bioassay development.
  • Provided as frozen vials with ≥5 x 10^6 cells per vial.
  • Surface expression confirmed by flow cytometry.
  • Suited for cell-based IL-1 RAcP binding and bioassay development.
  • Adherent CHO cell line with Zeocin selection (200 μg/mL).
  • Mycoplasma- and sterility-tested negative.
  • Validated passage stability and full traceable QC records.
  • Growth medium: F-12K + 10% FBS; freeze medium: serum-free cryopreservation medium.