Learn More
Genscript Corporation CHO-K1/4-1BB Stable Cell Line
Catalog No. 5000057467
Encompass
Recombinant CHO-K1 cells stably overexpress human TNF receptor superfamily member 9 (4-1BB) on the surface The surface expression of 4-1BB is validated by FACS analysis
Catalog No. 50-000-57467 Supplier Genscript Corporation Supplier No. M00538
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human TNF receptor superfamily member 9 (4-1BB) on the surface The surface expression of 4-1BB is validated by FACS analysis