Learn More
Genscript Corporation CHO-K1/5-HT1A/G15 Stable Cell
Catalog No. 5000057740
Encompass
The 5-Hydroxytryptamine receptor 1A (5-HT1A) is G sub i /sub -coupled GPCRs expressed in the cerebral cortex hippocampus septum amygdala and raphe nucelus with lesser amounts in the basal ganglia and thalamus Many functions of the central nervous system are influenced by 5-HT including sleep motor activity sensory perception arousal and appetite 5-HT1A ligands may prove to be therapeutic in the treatment of various disorders such as depression anxiety and schizophrenia
Catalog No. 50-000-57740 Supplier Genscript Corporation Supplier No. M00330
Add to Cart
Add to Cart
Request a Quote
The 5-Hydroxytryptamine receptor 1A (5-HT1A) is G sub i /sub -coupled GPCRs expressed in the cerebral cortex hippocampus septum amygdala and raphe nucelus with lesser amounts in the basal ganglia and thalamus Many functions of the central nervous system are influenced by 5-HT including sleep motor activity sensory perception arousal and appetite 5-HT1A ligands may prove to be therapeutic in the treatment of various disorders such as depression anxiety and schizophrenia