Learn More
Genscript Corporation CHO-K1/ACE2 Stable Cell Line
Catalog No. 5000056757
Encompass
Recombinant CHO-K1 cells stably express Angiotensin-converting enzyme 2 (ACE2) on the cell surface The surface expression of ACE2 is validated by FACS analysis This stable cell line product is designed for cell-based binding assays that measure binding affinity and stability of antibody based biologics binding with ACE2
Catalog No. 50-000-56757 Supplier Genscript Corporation Supplier No. M00771
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably express Angiotensin-converting enzyme 2 (ACE2) on the cell surface The surface expression of ACE2 is validated by FACS analysis This stable cell line product is designed for cell-based binding assays that measure binding affinity and stability of antibody based biologics binding with ACE2