Learn More
Genscript Corporation CHO-K1/ADORA1/G15 Stable Cell
Supplier:  Genscript Corporation M00324
Encompass
A1 is a receptor for adenosine It is demonstrated that the A1 receptor is ubiquitous throughout the entire body Activation of A1 elicits an inhibition of adenylate cyclase and therefore a decrease in the cAMP concentration A1 receptors are implicated in sleep promotion by inhibiting wake promoting cholinergic neurons in the basal forebrain Adenosine antagonists are widely used in neonatal medicine
Catalog No. 50-000-58381
Add to Cart
Request a Quote