Learn More
Genscript Corporation CHO-K1/ADORA2A/G15 Stable Cel
Catalog No. 5000058264
Encompass
Recombinant CHO-K1 cells stably overexpress human adenosine A2A receptor (ADORA2A) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58264 Supplier Genscript Corporation Supplier No. M00246
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human adenosine A2A receptor (ADORA2A) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways