Learn More
Genscript Corporation CHO-K1/ADORA2B/G15 Stable Cel
Supplier:  Genscript Corporation M00329
Encompass
Recombinant CHO-K1 cells stably overexpress human adenosine A2B receptor (ADORA2B) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57739
Add to Cart
Request a Quote