Learn More
Genscript Corporation CHO-K1/ADRA1A Stable Cell Line
Catalog No. 5000059042
Encompass
Recombinant CHO-K1 cells stably overexpress human adrenoceptor alpha 1A (ADRA1A) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-59042 Supplier Genscript Corporation Supplier No. M00225
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human adrenoceptor alpha 1A (ADRA1A) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways