Learn More
Genscript Corporation CHO-K1/ADRA1B Stable Cell Line
Catalog No. 5000058039
Encompass
Recombinant CHO-K1 cells stably overexpress human adrenoceptor alpha 1B (ADRA1B) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58039 Supplier Genscript Corporation Supplier No. M00260
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human adrenoceptor alpha 1B (ADRA1B) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways