Learn More
Genscript Corporation CHO-K1/ADRA2B/Gqi5 Stable Cell
Catalog No. 5000057443
Encompass
Adrenoceptors are 7-transmembrane receptors which mediate the central and peripheral actions of the neurotransmitter noradrenaline (norepinephrine) and the hormone and neurotransmitter adrenaline (epinephrine) Based on both pharmacological and molecular evidence they are divided into three major types - 1 2 and The 2 adrenoceptors include 3 subtypes - 2A 2B and 2C - each of which couples to G sub i/o /sub protein primarily br The 2B adrenoceptors (ADRA2B) is expressed in the kidney liver brain lung heart and skeletal muscle The development of 2B and 2C knock-out mice has shown that these two subtypes are not involved in the central hypotensive response to 2 agonists
Catalog No. 50-000-57443 Supplier Genscript Corporation Supplier No. M00454
Add to Cart
Add to Cart
Request a Quote
Adrenoceptors are 7-transmembrane receptors which mediate the central and peripheral actions of the neurotransmitter noradrenaline (norepinephrine) and the hormone and neurotransmitter adrenaline (epinephrine) Based on both pharmacological and molecular evidence they are divided into three major types - 1 2 and The 2 adrenoceptors include 3 subtypes - 2A 2B and 2C - each of which couples to G sub i/o /sub protein primarily br The 2B adrenoceptors (ADRA2B) is expressed in the kidney liver brain lung heart and skeletal muscle The development of 2B and 2C knock-out mice has shown that these two subtypes are not involved in the central hypotensive response to 2 agonists