Learn More
Genscript Corporation CHO-K1/B7-H2 Stable Cell Line
Catalog No. 5000057480
Encompass
Ligand for the T-cell-specific cell surface receptor ICOS acts as a costimulatory signal for T-cell proliferation and cytokine secretion It also induces B-cell proliferation and differentiation into plasma cells It could also play an important role in mediating local tissue responses to inflammatory conditions as well as modulating the secondary immune response by co-stimulating memory T-cell function
Catalog No. 50-000-57480 Supplier Genscript Corporation Supplier No. M00671
Add to Cart
Add to Cart
Request a Quote
Ligand for the T-cell-specific cell surface receptor ICOS acts as a costimulatory signal for T-cell proliferation and cytokine secretion It also induces B-cell proliferation and differentiation into plasma cells It could also play an important role in mediating local tissue responses to inflammatory conditions as well as modulating the secondary immune response by co-stimulating memory T-cell function