Learn More
Genscript Corporation CHO-K1/B7-H3 Stable Cell Line
Supplier:  Genscript Corporation M00536
Encompass
Recombinant CHO-K1 cells stably overexpress human CD276 (B7-H3) on the surface The surface expression of B7-H3 is validated by FACS analysis This cell line is recommended for cell-based binding assay to screen antibodies against B7-H3 or to measure binding affinity between B7-H3 and anti-B7-H3 antibodies
Catalog No. 50-000-57253
Only null left
Add to Cart
Request a Quote