Learn More
Genscript Corporation CHO-K1/BDCA2 and FcER1G Stable
Catalog No. 5000056732
Encompass
Recombinant CHO-K1 cells stably overexpress human C-type lectin domain family 4 member C (BDCA2) and Fc epsilon receptor Ig (FcER1G) on the surface The surface expression of BDCA2 is validated by FACS analysis
Catalog No. 50-000-56732 Supplier Genscript Corporation Supplier No. M00593
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human C-type lectin domain family 4 member C (BDCA2) and Fc epsilon receptor Ig (FcER1G) on the surface The surface expression of BDCA2 is validated by FACS analysis