Learn More
Genscript Corporation CHO-K1/BTLA Stable Cell Line
Catalog No. 5000058657
Encompass
The B and T lymphocyte attenuator (BTLA) is an Ig super family protein with an intermediate type Ig fold in the ectodomain and an ITIM inhibitory signaling domain in the cytosol BTLA interacts with the herpesvirus entry mediator (HVEM TNFRSF14) a TNFR super family member Engagement of BTLA by HVEM induces tyrosine phosphorylation of the ITIM motifs in the cytoplasmic tail of BTLA allowing the recruitment of the phosphatases SHP-1 and SHP-2 which attenuate signaling
Catalog No. 50-000-58657 Supplier Genscript Corporation Supplier No. M00534
Add to Cart
Add to Cart
Request a Quote
The B and T lymphocyte attenuator (BTLA) is an Ig super family protein with an intermediate type Ig fold in the ectodomain and an ITIM inhibitory signaling domain in the cytosol BTLA interacts with the herpesvirus entry mediator (HVEM TNFRSF14) a TNFR super family member Engagement of BTLA by HVEM induces tyrosine phosphorylation of the ITIM motifs in the cytoplasmic tail of BTLA allowing the recruitment of the phosphatases SHP-1 and SHP-2 which attenuate signaling