Learn More
Genscript Corporation CHO-K1/C5a/G15 Stable Cell Li
Catalog No. 5000057864
Encompass
The complement component 5a receptor 1 (C5a) is G sub q/11 /sub and G sub i/o /sub -coupled GPCRs C5a is a significant pathogenic driver in a number of immuno-inflammatory diseases making C5a inhibition an attractive therapeutic strategy C5a receptor plays an important role in mediating acute kidney allograft rejection
Catalog No. 50-000-57864 Supplier Genscript Corporation Supplier No. M00327
Add to Cart
Add to Cart
Request a Quote
The complement component 5a receptor 1 (C5a) is G sub q/11 /sub and G sub i/o /sub -coupled GPCRs C5a is a significant pathogenic driver in a number of immuno-inflammatory diseases making C5a inhibition an attractive therapeutic strategy C5a receptor plays an important role in mediating acute kidney allograft rejection