Learn More
Genscript Corporation CHO-K1/CB1/G15 Stable Cell Li
Catalog No. 5000059048
Encompass
The cannabinoid receptor CNR1 is Gs-coupled GPCRs expressed in primarily CNS and some peripheral neurones particularly prevalent in basal ganglia hippocampus cerebellum cerebral cortex and also presented in some non-neuronal cells and tissues for example leukocytes and testis A shorter human splice variant (411aa) has been identified in the brain and other tissues by reverse-transcriptase PCR but mRNA levels were less than 10-fold the levels of the longer isoform The pharmacological characteristics of the isoforms are similar
Catalog No. 50-000-59048 Supplier Genscript Corporation Supplier No. M00299
Add to Cart
Add to Cart
Request a Quote
The cannabinoid receptor CNR1 is Gs-coupled GPCRs expressed in primarily CNS and some peripheral neurones particularly prevalent in basal ganglia hippocampus cerebellum cerebral cortex and also presented in some non-neuronal cells and tissues for example leukocytes and testis A shorter human splice variant (411aa) has been identified in the brain and other tissues by reverse-transcriptase PCR but mRNA levels were less than 10-fold the levels of the longer isoform The pharmacological characteristics of the isoforms are similar