Learn More
Genscript Corporation CHO-K1/CB2 Stable Cell Line
Supplier:  Genscript Corporation M00433
Encompass
The Cannabinoid receptor 2 (CNR2) is a G sub i/o /sub -coupled GPCR expressed in spleen tonsils bone marrow and peripheral blood leukocytes There is one report that cannabinoid-induced inhibition of helper T cell activation is lost in macrophages obtained from CNR2 knockout mice CNR2 is a potential therapeutic target in the treatment of various disease conditions such as pain multiple sclerosis vascular disease Parkinson s disease and other central nerve system disorders
Catalog No. 50-000-57251
Add to Cart
Request a Quote