Learn More
Genscript Corporation CHO-K1/CD155 Stable Cell Line
Supplier:  Genscript Corporation M00591
Encompass
CD155 commonly known as PVR (poliovirus receptor) and Necl-5 (nectin-like molecule-5) is a type I transmembrane single-span glycoprotein and belongs to the nectins and nectin-like (Necl) subfamily CD155 was originally identified based on its ability to mediate the cell attachment and entry of poliovirus (PV) an etiologic agent of the central nervous system disease poliomyelitis The normal cellular function is in the establishment of intercellular adherens junctions between epithelial cells CD155 may assist in an efficient humoral immune response generated within the intestinal immune system It s been shown that CD155 can be recognized and bind by DNAM-1 and CD96 which promotes the adhension migration and NK-cell killing Thus inducing cell-mediated tumor-specific immunity
Catalog No. 50-000-57308
Add to Cart
Request a Quote