Learn More
Genscript Corporation CHO-K1/CD16A 158V Stable Cell
Catalog No. 5000059220
Encompass
Recombinant CHO-K1 cells stably overexpress human low affinity immunoglobulin gamma Fc region receptor III-A (Fc RIIIa/CD16A 158V) and Fc epsilon receptor Ig (FcER1G) on the surface The surface expression of CD16A 158V is validated by FACS analysis
Catalog No. 50-000-59220 Supplier Genscript Corporation Supplier No. M00597
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human low affinity immunoglobulin gamma Fc region receptor III-A (Fc RIIIa/CD16A 158V) and Fc epsilon receptor Ig (FcER1G) on the surface The surface expression of CD16A 158V is validated by FACS analysis