Learn More
Genscript Corporation CHO-K1/CD16B NA1 Stable Cell L
Catalog No. 5000057256
Encompass
Recombinant CHO-K1 cells stably overexpress human Fc fragment of IgG receptor IIIb (Fc RIIIb/CD16B NA1) The surface expression of CD16B NA1 is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies Fc fusion proteins or antibody based biologics that bind with CD16B during biologics research and development
Catalog No. 50-000-57256 Supplier Genscript Corporation Supplier No. M00602
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human Fc fragment of IgG receptor IIIb (Fc RIIIb/CD16B NA1) The surface expression of CD16B NA1 is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies Fc fusion proteins or antibody based biologics that bind with CD16B during biologics research and development