Learn More
Genscript Corporation CHO-K1/CD32A 131Arg Stable Cel
Catalog No. 5000056782
Encompass
Recombinant CHO-K1 cells stably overexpress human immunoglobulin gamma Fc region receptor II-a (Fc RIIa/CD32A 131Arg) on the surface The surface expression of CD32A 131Arg is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies and Fc fusion proteins or antibody based biologics binding with CD32A 131Arg GenScript also offers CD32A 131His stable cell line (Cat No M00598) for Fc RIIa polymorphism study
Catalog No. 50-000-56782 Supplier Genscript Corporation Supplier No. M00887
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human immunoglobulin gamma Fc region receptor II-a (Fc RIIa/CD32A 131Arg) on the surface The surface expression of CD32A 131Arg is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies and Fc fusion proteins or antibody based biologics binding with CD32A 131Arg GenScript also offers CD32A 131His stable cell line (Cat No M00598) for Fc RIIa polymorphism study