Learn More
Genscript Corporation CHO-K1/CD32B 232Thr Stable Cel
Catalog No. 5000058739
Encompass
Recombinant CHO-K1 cells stably overexpress human immunoglobulin gamma Fc region receptor II-b (Fc RIIb/CD32B 232Thr) on the surface The surface expression of CD32B 232Thr is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies and Fc fusion proteins or antibody based biologics binding with CD32B 232Thr GenScript also offers CD32B 232Ile stable cell line (Cat No M00587) for Fc RIIb polymorphism study
Catalog No. 50-000-58739 Supplier Genscript Corporation Supplier No. M00600
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human immunoglobulin gamma Fc region receptor II-b (Fc RIIb/CD32B 232Thr) on the surface The surface expression of CD32B 232Thr is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies and Fc fusion proteins or antibody based biologics binding with CD32B 232Thr GenScript also offers CD32B 232Ile stable cell line (Cat No M00587) for Fc RIIb polymorphism study