Learn More
Genscript Corporation CHO-K1/CD32B Ile232 Stable Cel
Supplier:  Genscript Corporation M00587
Encompass
Recombinant CHO-K1 cells stably overexpress human immunoglobulin gamma Fc region receptor II-b (Fc RIIb/CD32B 232Ile) on the surface The surface expression of CD32B 232Ile is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies and Fc fusion proteins or antibody based biologics binding with CD32B 232Ile GenScript also offers CD32B 232Thr stable cell line (Cat No M00600) for Fc RIIb polymorphism study
Catalog No. 50-000-58087
Add to Cart
Request a Quote