Learn More
Genscript Corporation CHO-K1/CD38 Stable Cell Line
Catalog No. 5000058056
Encompass
Recombinant CHO-K1 cells stably overexpress human CD38 on the surface The surface expression of CD38 is validated by FACS analysis
Catalog No. 50-000-58056 Supplier Genscript Corporation Supplier No. M00553
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human CD38 on the surface The surface expression of CD38 is validated by FACS analysis