Learn More
Genscript Corporation CHO-K1/CD64 Stable Cell Line
Catalog No. 5000057484
Encompass
Recombinant CHO-K1 cells stably overexpress high affinity human Fc fragment of IgG receptor Ia (Fc RI CD64) on the cell surface containing one alpha chain and two gamma chains The surface expression of CD64 is validated by FACS analysis This stable cell line product is designed for measuring the binding affinity and stability of the Fc region of antibodies Fc fusion proteins or antibody based biologics that bind with CD64 during biologics research and development
Catalog No. 50-000-57484 Supplier Genscript Corporation Supplier No. M00588
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress high affinity human Fc fragment of IgG receptor Ia (Fc RI CD64) on the cell surface containing one alpha chain and two gamma chains The surface expression of CD64 is validated by FACS analysis This stable cell line product is designed for measuring the binding affinity and stability of the Fc region of antibodies Fc fusion proteins or antibody based biologics that bind with CD64 during biologics research and development