Learn More
Genscript Corporation CHO-K1/CD80 Stable Cell Line
Catalog No. 5000058498
Encompass
Cluster of Differentiation 80 (also CD80 and B7-1) is a protein found on activated B cells and monocytes that provides a costimulatory signal necessary for T cell activation and survival It is the ligand for two different proteins on the T cell surface CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation) CD80 works in tandem with CD86 to prime T cells
Catalog No. 50-000-58498 Supplier Genscript Corporation Supplier No. M00545
Add to Cart
Add to Cart
Request a Quote
Cluster of Differentiation 80 (also CD80 and B7-1) is a protein found on activated B cells and monocytes that provides a costimulatory signal necessary for T cell activation and survival It is the ligand for two different proteins on the T cell surface CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation) CD80 works in tandem with CD86 to prime T cells