Learn More
Genscript Corporation CHO-K1/CMKLR1/G15 Stable Cell
Catalog No. 5000058394
Encompass
The orphan receptor ChemR23 (also referred to as CMKLR1 or DEZ) is a G-protein coupled receptor (GPCR) with homology to neuropeptide and chemoattractant receptors ChemR23 is also a G i-linked receptor expressed primarily by monocytes M s and plasmacytoid dendritic cells CMKLR1 has been shown to be expressed by DC generated in vitro from monocytes to mediate their migration to the proteolytically regulated chemoattractant chemerin and CMKLR1 may be a key mediator of pDC recruitment from the blood into tissue sites enriched in active chemerin
Catalog No. 50-000-58394 Supplier Genscript Corporation Supplier No. M00301
Add to Cart
Add to Cart
Request a Quote
The orphan receptor ChemR23 (also referred to as CMKLR1 or DEZ) is a G-protein coupled receptor (GPCR) with homology to neuropeptide and chemoattractant receptors ChemR23 is also a G i-linked receptor expressed primarily by monocytes M s and plasmacytoid dendritic cells CMKLR1 has been shown to be expressed by DC generated in vitro from monocytes to mediate their migration to the proteolytically regulated chemoattractant chemerin and CMKLR1 may be a key mediator of pDC recruitment from the blood into tissue sites enriched in active chemerin