Learn More
Genscript Corporation CHO-K1/CRE-CBLuc/GLP1R
Catalog No. 5000058519
Encompass
Recombinant CHO-K1 cells stably overexpress human glucagon like peptide 1 receptor (GLP1R) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58519 Supplier Genscript Corporation Supplier No. M00969
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human glucagon like peptide 1 receptor (GLP1R) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways