Learn More
Genscript Corporation CHO-K1/CRF1/G15 Stable Cell L
Supplier:  Genscript Corporation M00343
Encompass
Recombinant CHO-K1 cells stably overexpress human corticotropin releasing hormone receptor 1 (CRF1) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58640
Only null left
Add to Cart
Request a Quote