Learn More
Genscript Corporation CHO-K1/CRF2/G15 Stable Cell L
Catalog No. 5000057387
Encompass
Recombinant CHO-K1 cells stably overexpress human corticotropin releasing hormone receptor 2 (CRF2) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57387 Supplier Genscript Corporation Supplier No. M00208
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human corticotropin releasing hormone receptor 2 (CRF2) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways