Learn More
Genscript Corporation CHO-K1/CT/G15 Stable Cell Lin
Catalog No. 5000059120
Encompass
Recombinant CHO-K1 cells stably overexpress human calcitonin receptor (CT) on the surface and contain high levels of G protein G q and G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-59120 Supplier Genscript Corporation Supplier No. M00320
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human calcitonin receptor (CT) on the surface and contain high levels of G protein G q and G s to couple with the receptor in downstream signaling pathways