Learn More
Genscript Corporation CHO-K1/CTLA4 Stable Cell Line
Catalog No. 5000057305
Encompass
CTLA4 or CTLA-4 (cytotoxic T-lymphocyte-associated protein 4) also known as CD152 (cluster of differentiation 152) is a protein receptor that functions as an immune checkpoint and downregulates immune responses CTLA4 is constitutively expressed in regulatory T cells but only upregulated in conventional T cells after activation a phenomenon which is particularly notable in cancers It acts as an off switch when bound to CD80 or CD86 on the surface of antigen-presenting cells br/ The CTLA-4 protein is encoded by the Ctla4 gene in mice and the CTLA4 gene in humans
Catalog No. 50-000-57305 Supplier Genscript Corporation Supplier No. M00530
Add to Cart
Add to Cart
Request a Quote
CTLA4 or CTLA-4 (cytotoxic T-lymphocyte-associated protein 4) also known as CD152 (cluster of differentiation 152) is a protein receptor that functions as an immune checkpoint and downregulates immune responses CTLA4 is constitutively expressed in regulatory T cells but only upregulated in conventional T cells after activation a phenomenon which is particularly notable in cancers It acts as an off switch when bound to CD80 or CD86 on the surface of antigen-presenting cells br/ The CTLA-4 protein is encoded by the Ctla4 gene in mice and the CTLA4 gene in humans