Learn More
Genscript Corporation CHO-K1/cyno 4-1BB Stable Cell
Catalog No. 5000056768
Encompass
4-1BB is a member of the tumor necrosis factor (TNF) receptor family Its alternative names are tumor necrosis factor receptor superfamily member 9 (TNFRSF9) CD137 and induced by lymphocyte activation (ILA) It is currently of interest to immunologists as a co-stimulatory immune checkpoint molecule 4-1BB can be expressed by activated T cells but to a larger extent on CD8 than on CD4 T cells In addition 4-1BB expression is found on dendritic cells follicular dendritic cells natural killer cells granulocytes and cells of blood vessel walls at sites of inflammation
Catalog No. 50-000-56768 Supplier Genscript Corporation Supplier No. M00569
Add to Cart
Add to Cart
Request a Quote
4-1BB is a member of the tumor necrosis factor (TNF) receptor family Its alternative names are tumor necrosis factor receptor superfamily member 9 (TNFRSF9) CD137 and induced by lymphocyte activation (ILA) It is currently of interest to immunologists as a co-stimulatory immune checkpoint molecule 4-1BB can be expressed by activated T cells but to a larger extent on CD8 than on CD4 T cells In addition 4-1BB expression is found on dendritic cells follicular dendritic cells natural killer cells granulocytes and cells of blood vessel walls at sites of inflammation