Learn More
Genscript Corporation CHO-K1/cyno BDCA2 and FcER1G S
Catalog No. 5000058659
Encompass
Recombinant CHO-K1 cells stably overexpress cynomolgus C-type lectin domain family 4 member C (BDCA2) and Fc epsilon receptor Ig (FcER1G) on the surface The surface expression of BDCA2 is validated by FACS analysis
Catalog No. 50-000-58659 Supplier Genscript Corporation Supplier No. M00594
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress cynomolgus C-type lectin domain family 4 member C (BDCA2) and Fc epsilon receptor Ig (FcER1G) on the surface The surface expression of BDCA2 is validated by FACS analysis