Learn More
Genscript Corporation CHO-K1/cyno BDCA2 Stable Cell
Catalog No. 5000058074
Encompass
BDCA-2 is a novel type II C-type lectin which shows 50 7% sequence identity at the amino acid level to its putative murine ortholog the murine dendritic cell associated C-type lectin 2 In addition to its antigen capturing function BDCA-2 can mediate potent inhibition of induction of IFN- / expression in PDCs Production of IFN- / in response to several different types of viruses bacteria CpG-DNA dsRNA and SLE serum is by far the most prominent feature of PDCs
Catalog No. 50-000-58074 Supplier Genscript Corporation Supplier No. M00596
Add to Cart
Add to Cart
Request a Quote
BDCA-2 is a novel type II C-type lectin which shows 50 7% sequence identity at the amino acid level to its putative murine ortholog the murine dendritic cell associated C-type lectin 2 In addition to its antigen capturing function BDCA-2 can mediate potent inhibition of induction of IFN- / expression in PDCs Production of IFN- / in response to several different types of viruses bacteria CpG-DNA dsRNA and SLE serum is by far the most prominent feature of PDCs