Learn More
Genscript Corporation CHO-K1/cyno CTLA4 Stable Cell
Catalog No. 5000056769
Encompass
CTLA4 or CTLA-4 (cytotoxic T-lymphocyte-associated protein 4) also known as CD152 (cluster of differentiation 152) is a protein receptor that functioning as an immune checkpoint downregulates immune responses CTLA4 is constitutively expressed in Tregs but only upregulated in conventional T cells after activation It acts as an
Catalog No. 50-000-56769 Supplier Genscript Corporation Supplier No. M00571
Add to Cart
Add to Cart
Request a Quote
CTLA4 or CTLA-4 (cytotoxic T-lymphocyte-associated protein 4) also known as CD152 (cluster of differentiation 152) is a protein receptor that functioning as an immune checkpoint downregulates immune responses CTLA4 is constitutively expressed in Tregs but only upregulated in conventional T cells after activation It acts as an