Learn More
Genscript Corporation CHO-K1/cyno PD-1 Stable Cell L
Catalog No. 5000057336
Encompass
Recombinant CHO-K1 cells stably overexpress cynomolgus programmed cell death protein 1 (PD-1) on the surface The surface expression of PD-1 is validated by FACS analysis
Catalog No. 50-000-57336 Supplier Genscript Corporation Supplier No. M00572
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress cynomolgus programmed cell death protein 1 (PD-1) on the surface The surface expression of PD-1 is validated by FACS analysis