Learn More
Genscript Corporation CHO-K1/cyno PD-L2 Stable Cell
Supplier:  Genscript Corporation M00629
Encompass
Programmed cell death 1 ligand 2 (PD-L2) is a protein that is encoded by the PDCD1LG2 gene in humans PDCD1LG2 has also been designated as CD273 (cluster of differentiation 273) Inhibitory molecules of the B7/CD28 family play a key role in the induction of immune tolerance in the tumor microenvironment The programmed death-1 receptor (PD-1) with its ligands PD-L1 and PD-L2 constitutes an important member of these inhibitory pathways PD-L2 expression was initially thought to be restricted to antigen-presenting cells such as macrophages and dendritic cells (DCs) However PD-L2 expression can be induced on a wide variety of other immune cells and nonimmune cells depending on micro environmental stimuli
Catalog No. 50-000-58660
Only null left
Add to Cart
Request a Quote