Learn More
Genscript Corporation CHO-K1/cyno TIGIT Stable Cell
Catalog No. 5000056741
Encompass
TIGIT (T cell immunoreceptor with Ig and ITIM domains) was recently characterized as an inhibitory receptor which is expressed mainly on NK Treg CD8 T and CD4 T cells TIGIT harbors an immunoglobulin tail tyrosine (ITT)-like phosphorylation motif and an ITIM (immunoreceptor tyrosine-based inhibition motif) in its cytoplasmic tail The poliovirus receptor (PVR or CD155) was identified as the physical ligand of TIGIT with high affinity and PVRL2 (Nectin2 or CD112) also binds to TIGIT with a weaker binding capacity TIGIT/PVR engagement suppresses T cell activation through IL-10 secretion mediated by dendritic cells Furthermore TIGIT also exerts an intrinsic inhibitory function to T cell activation
Catalog No. 50-000-56741 Supplier Genscript Corporation Supplier No. M00672
Add to Cart
Add to Cart
Request a Quote
TIGIT (T cell immunoreceptor with Ig and ITIM domains) was recently characterized as an inhibitory receptor which is expressed mainly on NK Treg CD8 T and CD4 T cells TIGIT harbors an immunoglobulin tail tyrosine (ITT)-like phosphorylation motif and an ITIM (immunoreceptor tyrosine-based inhibition motif) in its cytoplasmic tail The poliovirus receptor (PVR or CD155) was identified as the physical ligand of TIGIT with high affinity and PVRL2 (Nectin2 or CD112) also binds to TIGIT with a weaker binding capacity TIGIT/PVR engagement suppresses T cell activation through IL-10 secretion mediated by dendritic cells Furthermore TIGIT also exerts an intrinsic inhibitory function to T cell activation