Learn More
Genscript Corporation CHO-K1/D2s/G15 Stable Cell Li
Catalog No. 5000058266
Encompass
The short form of D2 (D2S) and the long form (D2L) are two isoforms that contribute differentially to dopamine signaling in both prefrontal cortex and striatum The D2 dopamine receptor short form (D2s) has been shown to stimulate phospholipase D (PLD) activity independent of the activation of phospholipase C (PLC) activity in GH4 derived cells stably transfected with the D2s receptor Agonist activation of D2s has been shown to mediate the inhibition of growth in the same cell line
Catalog No. 50-000-58266 Supplier Genscript Corporation Supplier No. M00274
Add to Cart
Add to Cart
Request a Quote
The short form of D2 (D2S) and the long form (D2L) are two isoforms that contribute differentially to dopamine signaling in both prefrontal cortex and striatum The D2 dopamine receptor short form (D2s) has been shown to stimulate phospholipase D (PLD) activity independent of the activation of phospholipase C (PLC) activity in GH4 derived cells stably transfected with the D2s receptor Agonist activation of D2s has been shown to mediate the inhibition of growth in the same cell line