Learn More
Genscript Corporation CHO-K1/D5/G15 Stable Cell Lin
Catalog No. 5000058380
Encompass
Dopamine is the predominant catecholamine neurotransmitter found in mammalian brain where it controls a variety of functions including locomotor activity cognition emotion positive reinforcement food intake and endocrine regulation It also plays multiple roles in the periphery as a modulator of cardiovascular function catecholamine release hormone secretion vascular tone renal function and gastrointestinal motility The dopamine receptor family consists of five members which are classified into two groups D1-like (D1 and D5) and D2-like (D2 D3 and D4) D5 receptor has a 10-fold higher affinity for dopamine than the D1 receptor
Catalog No. 50-000-58380 Supplier Genscript Corporation Supplier No. M00322
Add to Cart
Add to Cart
Request a Quote
Dopamine is the predominant catecholamine neurotransmitter found in mammalian brain where it controls a variety of functions including locomotor activity cognition emotion positive reinforcement food intake and endocrine regulation It also plays multiple roles in the periphery as a modulator of cardiovascular function catecholamine release hormone secretion vascular tone renal function and gastrointestinal motility The dopamine receptor family consists of five members which are classified into two groups D1-like (D1 and D5) and D2-like (D2 D3 and D4) D5 receptor has a 10-fold higher affinity for dopamine than the D1 receptor