Learn More
Genscript Corporation CHO-K1/DP1/G15 Stable Cell Li
Supplier:  Genscript Corporation M00453
Encompass
DP1 is a G sub s /sub -coupled GPCR expressed in the colon eye platelets eosinophils that bind to PGD2 and PGE2 DP1 can inhibit platelet aggregation and also inhibit leukotriene B4 and superoxide anion (O2-) release from human neutrophils It can relax smooth muscle and regulate eosinophil apoptosis DP receptor knockout OVA-challenged mice exhibit reduced Th2 cytokine levels and lymphocyte accumulation in the lung In addition they failed to produce asthmatic responses
Catalog No. 50-000-57252
Add to Cart
Request a Quote