Learn More
Genscript Corporation CHO-K1/DP2/G15 Stable Cell Li
Catalog No. 5000057301
Encompass
Prostaglandin D(2) (PGD(2)) is derived from arachidonic acid and binds with high affinity to the G-protein coupled receptors prostanoid DP(1) and DP(2) Interaction with DP(2) results in cell chemotaxis eosinophil degranulation eosinophil shape change adhesion molecule upregulation and Th2 cytokine production In allergic rhinitis and allergic asthma PGD(2) is released from mast cells in response to allergen challenge and may trigger symptoms such as sneezing rhinorrhea pruritus mucus hypersecretion and pulmonary inflammation GenScript s cloned human DP2-expressing cell line co-expressing G 15 is made in the CHO-K1 host
Catalog No. 50-000-57301 Supplier Genscript Corporation Supplier No. M00436
Add to Cart
Add to Cart
Request a Quote
Prostaglandin D(2) (PGD(2)) is derived from arachidonic acid and binds with high affinity to the G-protein coupled receptors prostanoid DP(1) and DP(2) Interaction with DP(2) results in cell chemotaxis eosinophil degranulation eosinophil shape change adhesion molecule upregulation and Th2 cytokine production In allergic rhinitis and allergic asthma PGD(2) is released from mast cells in response to allergen challenge and may trigger symptoms such as sneezing rhinorrhea pruritus mucus hypersecretion and pulmonary inflammation GenScript s cloned human DP2-expressing cell line co-expressing G 15 is made in the CHO-K1 host