Learn More
Genscript Corporation CHO-K1/EDNRB Stable Cell Line
Supplier:  Genscript Corporation M00150
Encompass
In mammals the endothelin (ET) family comprises three endogenous isoforms ET-1 ET-2 and ET-3 These endothelium-derived peptides perform their functions via two endothelin receptors classified as EDNRA and EDNRB EDNRB receptors present on the endothelial cells lining the vessel wall and play a role in the release of endothelium-derived relaxing factors such as nitric oxide (NO) and prostanoids and in removing ET from the circulation
Catalog No. 50-000-59043
Add to Cart
Request a Quote