Learn More
Genscript Corporation CHO-K1/EP2/G15 Stable Cell Li
Catalog No. 5000058269
Encompass
Prostaglandin (PG) E2 exerts its actions by acting on a group of G-protein-coupled receptors (GPCRs) There are four GPCRs responding to PGE2 designated subtypes EP1 EP2 EP3 and EP4 and multiple splicing isoforms of the subtype EP3 The EP subtypes exhibit differences in signal transduction tissue localization and expression regulation Studies have identified that EP2 mediates many processes such as facilitating ovulation and fertilization suppressing dendritic cell differentiation and promoting amyloid- formation in Alzheimer s disease
Catalog No. 50-000-58269 Supplier Genscript Corporation Supplier No. M00311
Add to Cart
Add to Cart
Request a Quote
Prostaglandin (PG) E2 exerts its actions by acting on a group of G-protein-coupled receptors (GPCRs) There are four GPCRs responding to PGE2 designated subtypes EP1 EP2 EP3 and EP4 and multiple splicing isoforms of the subtype EP3 The EP subtypes exhibit differences in signal transduction tissue localization and expression regulation Studies have identified that EP2 mediates many processes such as facilitating ovulation and fertilization suppressing dendritic cell differentiation and promoting amyloid- formation in Alzheimer s disease